accessibility menu, dialog, popup

UserName

CAS RN 4488-57-7

IUPAC Name: tricyclo[5.2.1.0²,⁶]dec-3-ene
Synonyms: hexahydro-methanoindene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: HANKSFAYJLDDKP-UHFFFAOYNA-N
SMILES: C1CC2CC1C1CC=CC21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results