accessibility menu, dialog, popup

UserName

CH3OO(S)HONH2

CAS RN 4465-44-5

IUPAC Name: [(2S)-3-hydroxy-1-methoxy-1-oxopropan-2-yl](triphenylmethyl)azanium
Synonyms: [(2S)-3-hydroxy-1-methoxy-1-oxopropan-2-yl](triphenylmethyl)azanium
Molecular Weight (g/mol): 362.45
Molecular Formula: C23H24NO3
InChi Key: LXAWQKKSNNYYEK-NRFANRHFSA-O
SMILES: COC(=O)[C@H](CO)[NH2+]C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results