accessibility menu, dialog, popup

UserName

HOOF

CAS RN 445-29-4

IUPAC Name: 2-fluorobenzoic acid
Synonyms: 2-fluorobenzoic acid o-fluorbenzoesaeure
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: NSTREUWFTAOOKS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1F
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)