CAS RN 445-28-3
CAS RN 445-28-3
IUPAC Name:
2-fluorobenzamide
Synonyms:
2-fluorobenzamide
Molecular Weight (g/mol):
139.13
Molecular Formula:
C7H6FNO
InChi Key:
KGGHWIKBOIQEAJ-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=CC=C1F
1
–
1
of
1
results
1
2-Fluorobenzamide 98.0+%, TCI America™
CAS: 445-28-3 Molecular Formula: C7H6FNO Molecular Weight (g/mol): 139.129 MDL Number: MFCD00007970 InChI Key: KGGHWIKBOIQEAJ-UHFFFAOYSA-N Synonym: o-fluorobenzamide,benzamide, 2-fluoro,benzamide, o-fluoro,o-fluorobenzoic acid amide,ortho-fluorobenzamide,fluorobenzamide,2-fluoro-benzamide,pubchem3405,acmc-1aljb,2-fluorobenzamide PubChem CID: 67964 IUPAC Name: 2-fluorobenzamide SMILES: C1=CC=C(C(=C1)C(=O)N)F