accessibility menu, dialog, popup

UserName

CAS RN 445-26-1

H3COHF

CAS RN 445-26-1

IUPAC Name: 1-(2-fluorophenyl)ethan-1-ol
Synonyms: 1-(2-fluorophenyl)ethanol
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: SXFYVXSOEBCFLV-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results