accessibility menu, dialog, popup

UserName

HOOOO

CAS RN 4442-53-9

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
Synonyms: 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: VCLSWKVAHAJSFL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=C1OCCO2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results