accessibility menu, dialog, popup

UserName

H3CBOHOH

CAS RN 4426-47-5

IUPAC Name: butylboronic acid
Synonyms: 1-butane boronic acid
Molecular Weight (g/mol): 101.94
Molecular Formula: C4H11BO2
InChi Key: QPKFVRWIISEVCW-UHFFFAOYSA-N
SMILES: CCCCB(O)O
Quantity 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)