accessibility menu, dialog, popup

UserName

HClH2NNHNH2S

CAS RN 4346-94-5

IUPAC Name: hydrogen aminothiourea chloride
Synonyms: hydrogen thiosemicarbazide chloride
Molecular Weight (g/mol): 127.59
Molecular Formula: CH6ClN3S
InChi Key: LEHOGBUBQSGCOK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC(N)=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results