missing translation for 'onlineSavingsMsg'
Learn More

Thiosemicarbazide Hydrochloride 98.0+%, TCI America™

Catalog No. T022225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T022225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T022225G Supplier TCI America Supplier No. T022225G
Only null left

Specifications

CAS 4346-94-5
Melting Point 171°C
Color White
Molecular Formula CH6ClN3S
Quantity 25 g
MDL Number MFCD00060206
UN Number 2811
InChI Key LEHOGBUBQSGCOK-UHFFFAOYSA-N
SMILES C(=S)(N)NN.Cl
IUPAC Name aminothiourea;hydrochloride
Molecular Weight (g/mol) 127.59
PubChem CID 3037943
Formula Weight 127.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiosemicarbazide Hydrochloride
Show More Show Less
RTECSNumber VT4300000
TSCA No