accessibility menu, dialog, popup

UserName

CAS RN 4337-04-6

IUPAC Name: 2-[4-(butanoyloxy)-3-hydroxy-5-oxo-2,5-dihydrofuran-2-yl]-2-hydroxyethyl butanoate
Synonyms: 2-[4-(butanoyloxy)-3-hydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl butanoate
Molecular Weight (g/mol): 316.31
Molecular Formula: C14H20O8
InChi Key: BFXWCTCPRAYDEB-UHFFFAOYNA-N
SMILES: CCCC(=O)OCC(O)C1OC(=O)C(OC(=O)CCC)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results