accessibility menu, dialog, popup

UserName

CH3SNH3C

CAS RN 4293-61-2

IUPAC Name: 4-ethyl-2-methyl-4,5-dihydro-1,3-thiazole
Synonyms: 4-ethyl-2-methyl-4,5-dihydro-1,3-thiazole
Molecular Weight (g/mol): 129.22
Molecular Formula: C6H11NS
InChi Key: LENVRVUQOXYSDH-UHFFFAOYNA-N
SMILES: CCC1CSC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results