accessibility menu, dialog, popup

UserName

OHOHNO

CAS RN 4289-95-6

IUPAC Name: 2-formamido-3-phenylpropanoic acid
Synonyms: 2-formamido-3-phenylpropanoic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: NSTPXGARCQOSAU-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results