accessibility menu, dialog, popup

UserName

CAS RN 42831-50-5

CH3ONOO

CAS RN 42831-50-5

IUPAC Name: 5-methyl-1,2-oxazole-4-carboxylate
Synonyms: 5-methyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol): 126.09
Molecular Formula: C5H4NO3
InChi Key: VQBXUKGMJCPBMF-UHFFFAOYSA-M
SMILES: CC1=C(C=NO1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results