CAS RN 42771-79-9
CAS RN 42771-79-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Acetyl-2-Fluorobiphenyl, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.