accessibility menu, dialog, popup

UserName

H3CNCH3SOOO

CAS RN 4272-77-9

IUPAC Name: 5-(dimethylamino)naphthalene-1-sulfonate
Synonyms: 5-(dimethylamino)naphthalene-1-sulfonate
Molecular Weight (g/mol): 250.29
Molecular Formula: C12H12NO3S
InChi Key: BBEQQKBWUHCIOU-UHFFFAOYSA-M
SMILES: CN(C)C1=C2C=CC=C(C2=CC=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results