accessibility menu, dialog, popup

UserName

CH3NOHN

CAS RN 4241-27-4

IUPAC Name: 6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Synonyms: 6-methyl-2-oxo-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: FIMGYEKEYXUTGD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C#N)C(=O)N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)