accessibility menu, dialog, popup

UserName

CH3NCH3NH2

CAS RN 42389-54-8

IUPAC Name: N1,N1-diethylcyclohexane-1,4-diamine
Synonyms: N1,N1-diethylcyclohexane-1,4-diamine
Molecular Weight (g/mol): 170.3
Molecular Formula: C10H22N2
InChi Key: ZFWPGRUTHAZDBK-UHFFFAOYSA-N
SMILES: CCN(CC)C1CCC(N)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results