accessibility menu, dialog, popup

UserName

NH2SOOFFF

CAS RN 421-85-2

IUPAC Name: trifluoromethanesulfonamide
Synonyms: trifluoromethanesulfonamide
Molecular Weight (g/mol): 149.09
Molecular Formula: CH2F3NO2S
InChi Key: KAKQVSNHTBLJCH-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)