CAS RN 42074-68-0
IUPAC Name:
1-chloro-2-(chlorodiphenylmethyl)benzene
Synonyms:
1-chloro-2-(chlorodiphenylmethyl)benzene
Molecular Weight (g/mol):
313.22
Molecular Formula:
C19H14Cl2
InChi Key:
JFLSOKIMYBSASW-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1C(Cl)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g
CAS: 42074-68-0 Molecular Formula: C19H14Cl2 Molecular Weight (g/mol): 313.221 MDL Number: MFCD00040399 InChI Key: JFLSOKIMYBSASW-UHFFFAOYSA-N Synonym: 2-chlorotrityl chloride,chloro 2-chlorophenyl methylene dibenzene,1-chloro-2-chlorodiphenylmethyl benzene,2-chlorophenyldiphenylmethyl chloride,2-chlorotrityl chloride resin,2-chlorophenyl diphenylmethyl chloride,2-chlorotrityl resin,2-chlorotritylchloride,chloro 2-chlorophenyl diphenylmethane,2-chlorotrityl chloride, polymer-bound PubChem CID: 94524 IUPAC Name: 1-chloro-2-[chloro(diphenyl)methyl]benzene SMILES: C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)Cl