accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 42059-81-4

IUPAC Name: 6-methyl-4-oxo-4H-chromene-3-carbaldehyde
Synonyms: 6-methyl-4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: GBWMIOYSMWCYIZ-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC=C(C=O)C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results