missing translation for 'onlineSavingsMsg'
Learn More

4-(9H-Carbazol-9-yl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C29261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29261G Supplier TCI America Supplier No. C29261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-(9H-Carbazol-9-yl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 419536-33-7
Melting Point 264°C
Color White
Molecular Formula C18H14BNO2
Quantity 1 g
MDL Number MFCD13176534
Synonym 4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
InChI Key JGAVTCVHDMOQTJ-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
IUPAC Name [4-(9H-carbazol-9-yl)phenyl]boronic acid
Molecular Weight (g/mol) 287.13
PubChem CID 23120827
Formula Weight 287.13
Physical Form Crystalline Powder
Show More Show Less