accessibility menu, dialog, popup

UserName

CAS RN 4186-54-3

IUPAC Name: 2,4-dibromo-6-(bromomethyl)phenol
Synonyms: 2,4-dibromo-6-(bromomethyl)phenol
Molecular Weight (g/mol): 344.83
Molecular Formula: C7H5Br3O
InChi Key: SPUZPJKEJXIRSF-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Br)C=C1CBr
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results