accessibility menu, dialog, popup

UserName

CAS RN 4185-69-7

ClONONOON

CAS RN 4185-69-7

IUPAC Name: 1-(2,4-dinitrophenyl)-1λ⁵-pyridin-1-ylium chloride
Synonyms: 1-(2,4-dinitrophenyl)-1λ⁵-pyridin-1-ylium chloride
Molecular Weight (g/mol): 281.65
Molecular Formula: C11H8ClN3O4
InChi Key: UYHMQTNGMUDVIY-UHFFFAOYSA-M
SMILES: [Cl-].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results