CAS RN 41830-80-2
IUPAC Name:
12-amino-8-(dimethylamino)-7-methyl-10λ⁴-oxa-5-azatetraphen-10-ylium chloride
Synonyms:
12-amino-8-(dimethylamino)-7-methyl-10λ⁴-oxa-5-azatetraphen-10-ylium chloride
Molecular Weight (g/mol):
339.82
Molecular Formula:
C19H18ClN3O
InChi Key:
ABASSERKNXVFAX-UHFFFAOYSA-M
SMILES:
[Cl-].CN(C)C1=CC2=[O+]C3=C(N=C2C=C1C)C1=CC=CC=C1C(N)=C3
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Cresyl Violet perchlorate, 98%, pure
Thermo Scientific Chemicals Cresyl Violet perchlorate, 98%, pure
CAS: 41830-80-2 Molecular Formula: C16H12ClN3O5 Molecular Weight (g/mol): 361.738 MDL Number: MFCD00012671 InChI Key: HLHUTAPXZITNPJ-UHFFFAOYSA-N Synonym: Oxazine 9 perchlorate PubChem CID: 621529 IUPAC Name: 5-iminobenzo[a]phenoxazin-9-amine;perchloric acid SMILES: C1=CC=C2C(=C1)C(=N)C=C3C2=NC4=C(O3)C=C(C=C4)N.OCl(=O)(=O)=O