accessibility menu, dialog, popup

UserName

CAS RN 4139-61-1

BrOOOH

CAS RN 4139-61-1

IUPAC Name: 6-bromo-4-hydroxy-2H-chromen-2-one
Synonyms: 6-bromo-4-hydroxychromen-2-one
Molecular Weight (g/mol): 241.04
Molecular Formula: C9H5BrO3
InChi Key: KNMCTCABMSGXGR-UHFFFAOYSA-N
SMILES: OC1=CC(=O)OC2=CC=C(Br)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results