accessibility menu, dialog, popup

UserName

HClCH3NH3COO

CAS RN 4136-37-2

IUPAC Name: hydrogen 1,1-dimethylpyrrolidin-1-ium-2-carboxylate chloride
Synonyms: hydrogen 1,1-dimethylpyrrolidin-1-ium-2-carboxylate chloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: DUNMULOWUUIQIL-UHFFFAOYNA-N
SMILES: [H+].[Cl-].C[N+]1(C)CCCC1C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results