CAS RN 4131-74-2
IUPAC Name:
methyl 3-[(3-methoxy-3-oxopropyl)sulfanyl]propanoate
Synonyms:
methyl 3-[(3-methoxy-3-oxopropyl)sulfanyl]propanoate
Molecular Weight (g/mol):
206.26
Molecular Formula:
C8H14O4S
InChi Key:
MYWWWNVEZBAKHR-UHFFFAOYSA-N
SMILES:
COC(=O)CCSCCC(=O)OC
1
–
1
of
1
results
1
Dimethyl 3,3'-Thiodipropionate 98.0+%, TCI America™
CAS: 4131-74-2 Molecular Formula: C8H14O4S Molecular Weight (g/mol): 206.256 MDL Number: MFCD00008465 InChI Key: MYWWWNVEZBAKHR-UHFFFAOYSA-N Synonym: 3,3′C-Thiodipropionic Acid Dimethyl Ester PubChem CID: 77781 IUPAC Name: methyl 3-(3-methoxy-3-oxopropyl)sulfanylpropanoate SMILES: COC(=O)CCSCCC(=O)OC