accessibility menu, dialog, popup

UserName

H2COH

CAS RN 4117-10-6

IUPAC Name: hept-6-en-1-ol
Synonyms: hept-6-en-1-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: UFULDTPDHIRNGS-UHFFFAOYSA-N
SMILES: OCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results