accessibility menu, dialog, popup

UserName

CH3SOONSO

CAS RN 4104-47-6

IUPAC Name: 4-methyl-N-(oxo-λ⁴-sulfanylidene)benzene-1-sulfonamide
Synonyms: 4-methyl-N-(oxo-λ⁴-sulfanylidene)benzenesulfonamide
Molecular Weight (g/mol): 217.26
Molecular Formula: C7H7NO3S2
InChi Key: VKTSIMMJOIPMGE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N=S=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results