accessibility menu, dialog, popup

UserName

CAS RN 4100-41-8

IUPAC Name: 1,2,3-thiadiazol-5-amine
Synonyms: 1,2,3-thiadiazol-5-amine
Molecular Weight (g/mol): 101.13
Molecular Formula: C2H3N3S
InChi Key: PVGHNTXQMCYYGF-UHFFFAOYSA-N
SMILES: NC1=CN=NS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results