CAS RN 40957-83-3
IUPAC Name:
7-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one
Synonyms:
glycitein
Molecular Weight (g/mol):
284.27
Molecular Formula:
C16H12O5
InChi Key:
DXYUAIFZCFRPTH-UHFFFAOYSA-N
SMILES:
COC1=C(O)C=C2OC=C(C(=O)C2=C1)C1=CC=C(O)C=C1
1
–
2
of
2
results
1
phyproof™ Glycitein, ≥95% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
2
Glycitein
CAS: 40957-83-3 Molecular Formula: C16H12O5 Molecular Weight (g/mol): 284.27 InChI Key: DXYUAIFZCFRPTH-UHFFFAOYSA-N IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one SMILES: COC1=C(O)C=C2OC=C(C(=O)C2=C1)C1=CC=C(O)C=C1