accessibility menu, dialog, popup

UserName

H3CCH3SOH

CAS RN 40811-49-2

IUPAC Name: 2-(propan-2-ylsulfanyl)ethan-1-ol
Synonyms: 2-(isopropylsulfanyl)ethanol
Molecular Weight (g/mol): 120.21
Molecular Formula: C5H12OS
InChi Key: QGONIKZRWOOHBB-UHFFFAOYSA-N
SMILES: CC(C)SCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results