CAS RN 4074-25-3
CAS RN 4074-25-3
IUPAC Name:
1,3,5-tri-tert-butyl-2-nitrobenzene
Synonyms:
1,3,5-tri-tert-butyl-2-nitrobenzene
Molecular Weight (g/mol):
291.44
Molecular Formula:
C18H29NO2
InChi Key:
IMDZOFHRUMJNQR-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)[N+]([O-])=O)C(C)(C)C
1
–
1
of
1
results
1
2,4,6-Tri-tert-butylnitrobenzene 95.0+%, TCI America™
CAS: 4074-25-3 Molecular Formula: C18H29NO2 Molecular Weight (g/mol): 291.435 MDL Number: MFCD00008818 InChI Key: IMDZOFHRUMJNQR-UHFFFAOYSA-N Synonym: 2,4,6-tri-tert-butylnitrobenzene,2,4,6-tri-t-butylnitrobenzene,1,3,5-tri-tert-butyl-2-nitrobenzene,1,3,5-tri tert-butyl-2-nitrobenzene,2,4,6 tri-tert-butylnitrobenzene,2-nitro-1,3,5-tri-tert-butylbenzene,acmc-209jgh,1,3,5-tritert-butyl-2-nitro-benzene,2-nitro-1,3,5-tritert-butyl-benzene,2,4,6-tri-tert-butyl-1-nitrobenzene PubChem CID: 77691 IUPAC Name: 1,3,5-tritert-butyl-2-nitrobenzene SMILES: CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)[N+](=O)[O-])C(C)(C)C