accessibility menu, dialog, popup

UserName

CAS RN 4043-59-8

H3COOHOCH3

CAS RN 4043-59-8

IUPAC Name: 1,3-diethoxypropan-2-ol
Synonyms: 1,3-diethoxypropan-2-ol
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: WIHIUTUAHOZVLE-UHFFFAOYSA-N
SMILES: CCOCC(O)COCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results