accessibility menu, dialog, popup

UserName

CAS RN 40393-99-5

IUPAC Name: (2,4,6-trimethylphenyl)methanaminium
Synonyms: (2,4,6-trimethylphenyl)methanaminium
Molecular Weight (g/mol): 150.24
Molecular Formula: C10H16N
InChi Key: DGSRAILDFBJNQI-UHFFFAOYSA-O
SMILES: CC1=CC(C)=C(C[NH3+])C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results