accessibility menu, dialog, popup

UserName

CAS RN 40191-32-0

IUPAC Name: oxane-4-carbonyl chloride
Synonyms: oxane-4-carbonyl chloride
Molecular Weight (g/mol): 148.59
Molecular Formula: C6H9ClO2
InChi Key: RYGUCYSSMOFTSH-UHFFFAOYSA-N
SMILES: ClC(=O)C1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results