accessibility menu, dialog, popup

UserName

CAS RN 40101-31-3

BrNOO

CAS RN 40101-31-3

IUPAC Name: 2-(4-bromophenyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(4-bromophenyl)isoindole-1,3-dione
Molecular Weight (g/mol): 302.13
Molecular Formula: C14H8BrNO2
InChi Key: DNQCPYWSXMATIN-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results