accessibility menu, dialog, popup

UserName

BOHOH

CAS RN 400607-31-0

IUPAC Name: (9,9-diphenyl-9H-fluoren-2-yl)boronic acid
Synonyms: 9,9-diphenylfluoren-2-ylboronic acid
Molecular Weight (g/mol): 362.24
Molecular Formula: C25H19BO2
InChi Key: CPFALCJMNUHBHP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results