CAS RN 3987-92-6
IUPAC Name:
methyl 4-amino-3-nitrobenzoate
Synonyms:
methyl 4-amino-3-nitrobenzoate
Molecular Weight (g/mol):
196.16
Molecular Formula:
C8H8N2O4
InChi Key:
HNTLUEZVPLRQEV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(=C(N)C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Methyl 4-amino-3-nitrobenzoate, 97%
Methyl 4-amino-3-nitrobenzoate, 97%
CAS: 3987-92-6 Molecular Formula: C8H8N2O4 Molecular Weight (g/mol): 196.162 MDL Number: MFCD00017562 InChI Key: HNTLUEZVPLRQEV-UHFFFAOYSA-N Synonym: methyl 4-amino-3-nitrobenzenecarboxylate,benzoic acid, 4-amino-3-nitro-, methyl ester,4-amino-3-nitro-benzoic acid methyl ester,4-methoxycarbonyl-2-nitroaniline,4-amino-3-nitromethylbenzoate,methyl 3-nitro-4-aminobenzoate,2-amino-5-methoxycarbonyl nitrobenzene,3-nitro-4-aminobenzoic acid methyl ester,4-amino-3-nitrobenzoicacidmethylester,acmc-209j8j PubChem CID: 3941008 IUPAC Name: methyl 4-amino-3-nitrobenzoate SMILES: COC(=O)C1=CC(=C(C=C1)N)[N+](=O)[O-]