CAS RN 3968-92-1
IUPAC Name:
(butan-2-yl)triphenylphosphanium bromide
Synonyms:
triphenyl(sec-butyl)phosphanium bromide
Molecular Weight (g/mol):
399.31
Molecular Formula:
C22H24BrP
InChi Key:
HKWAYXKPYJOYEL-UHFFFAOYNA-M
SMILES:
[Br-].CCC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
(2-Butyl)triphenylphosphonium bromide, 96%
(2-Butyl)triphenylphosphonium bromide, 96%
CAS: 3968-92-1 Molecular Formula: C22H24BrP Molecular Weight (g/mol): 399.31 MDL Number: MFCD00050236 InChI Key: HKWAYXKPYJOYEL-UHFFFAOYNA-M Synonym: 2-butyl triphenylphosphonium bromide,sec-butyltriphenylphosphonium bromide,triphenyl sec-butyl phosphanium bromide,acmc-1cswd,butan-2-yl triphenyl phosphanium bromide,phosphonium, 1-methylpropyl triphenyl-, bromide 1:1 PubChem CID: 3083826 IUPAC Name: butan-2-yl(triphenyl)phosphanium;bromide SMILES: [Br-].CCC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1