CAS RN 3966-30-1
CAS RN 3966-30-1
IUPAC Name:
2-hydroxy-2-phenylpropanoic acid
Synonyms:
(+-)-atrolactic acid (1)-2-phenyllactic acid hydroxymethylmandelic acid atrolactic acid, (+-)- atrolactinic acid dl-atrolactic acid (+-)-isomer atrolactic acid dl-atrolactate (S)-atrolactic acid atrolactinate
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
NWCHELUCVWSRRS-UHFFFAOYNA-N
SMILES:
CC(O)(C(O)=O)C1=CC=CC=C1
1
–
1
of
1
results
1
(R)-(-)-2-Hydroxy-2-phenylpropionic acid, 98+%, Thermo Scientific Chemicals
CAS: 3966-30-1 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.176 MDL Number: MFCD00067699 InChI Key: NWCHELUCVWSRRS-SECBINFHSA-N Synonym: r---2-hydroxy-2-phenylpropionic acid,2r-2-hydroxy-2-phenylpropanoic acid,unii-h38dkr9931,r-2-hydroxy-2-phenylpropanoic acid,phenyllactic acid,atrolactic acid,-,2-hydroxy-2-phenylpropanoic acid #,r---atrolactic acid,r-phenyl lactic acid,r-alpha-hydroxy-alpha-methylbenzeneacetic acid PubChem CID: 736858 IUPAC Name: (2R)-2-hydroxy-2-phenylpropanoic acid SMILES: CC(C1=CC=CC=C1)(C(=O)O)O