accessibility menu, dialog, popup

UserName

CAS RN 39216-86-9

IUPAC Name: 4-(ethylamino)butan-1-ol
Synonyms: 4-(ethylamino)butan-1-ol
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: PVNNOLUAMRODAC-UHFFFAOYSA-N
SMILES: CCNCCCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results