CAS RN 38997-97-6
IUPAC Name:
1-(2,3-dihydro-1H-inden-4-yl)ethan-1-one
Synonyms:
1-(2,3-dihydro-1H-inden-4-yl)ethanone
Molecular Weight (g/mol):
160.22
Molecular Formula:
C11H12O
InChi Key:
FPHUGFGBPNVMQR-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C2CCCC2=CC=C1
1
–
1
of
1
results
1
4-Acetylindan 98.0+%, TCI America™
CAS: 38997-97-6 Molecular Formula: C11H12O Molecular Weight (g/mol): 160.216 MDL Number: MFCD09033715 InChI Key: FPHUGFGBPNVMQR-UHFFFAOYSA-N PubChem CID: 13657961 IUPAC Name: 1-(2,3-dihydro-1H-inden-4-yl)ethanone SMILES: CC(=O)C1=C2CCCC2=CC=C1