accessibility menu, dialog, popup

UserName

CAS RN 38769-92-5

IUPAC Name: chloro[(4-methoxyphenyl)methyl]magnesium
Synonyms: chloro[(4-methoxyphenyl)methyl]magnesium
Molecular Weight (g/mol): 180.91
Molecular Formula: C8H9ClMgO
InChi Key: XHDYUCUVUZSISM-UHFFFAOYSA-M
SMILES: COC1=CC=C(C[Mg]Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results