accessibility menu, dialog, popup

UserName

CH3ONH3CCH3OCH3

CAS RN 38711-20-5

IUPAC Name: (2,2-dimethoxyethyl)dimethylamine
Synonyms: (2,2-dimethoxyethyl)dimethylamine
Molecular Weight (g/mol): 133.19
Molecular Formula: C6H15NO2
InChi Key: HUYAEQCJNXODLQ-UHFFFAOYSA-N
SMILES: COC(CN(C)C)OC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)