missing translation for 'onlineSavingsMsg'
Learn More

(Dimethylamino)acetaldehyde Dimethyl Acetal 98.0+%, TCI America™

Catalog No. D170425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D170425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D170425ML Supplier TCI America Supplier No. D170425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Synthetic building block for chemical research.

Specifications

CAS 38711-20-5
Color Yellow
Boiling Point 81°C
Molecular Formula C6H15NO2
Quantity 25 mL
MDL Number MFCD00010205
UN Number 1993
Synonym ethanamine, 2,2-dimethoxy-n,n-dimethyl, dimethylamino acetaldehyde dimethyl acetal, dimethylaminoacetaldehyde dimethyl acetal, dimethylaminoacetaldehyde dimethylacetal, n-2,2-dimethoxyethyl dimethylamine, 2-dimethylamino acetaldehyde dimethylacetal, 2,2-dimethoxyethyl dimethylamine, 2,2-dimethoxyethyl dimethyl amine, 2,2-dimethoxy-n,n-dimethylethanamin, acmc-209j1l
InChI Key HUYAEQCJNXODLQ-UHFFFAOYSA-N
SMILES COC(CN(C)C)OC
IUPAC Name (2,2-dimethoxyethyl)dimethylamine
Molecular Weight (g/mol) 133.19
PubChem CID 123476
Formula Weight 133.19
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material (Dimethylamino)acetaldehyde Dimethyl Acetal
Show More Show Less
EINECSNumber (2)-0530
TSCA Yes