accessibility menu, dialog, popup

UserName

CAS RN 38562-01-5

H3C(S)OH(R)(R)HO(S)OH(R)OHONH2HOOHHO(E)(Z)

CAS RN 38562-01-5

IUPAC Name: (5Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxyoct-1-en-1-yl]cyclopentyl]hept-5-enoic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol
Synonyms: dinoprost tromethamine PGF2-α tham PGF2α tham prostaglandin F2α tham dinoprost, trometamol salt dinolytic prostin F2 α prostalmon F panacelan F tromethamine salt prostaglandin F2a tromethamine
Molecular Weight (g/mol): 475.62
Molecular Formula: C24H45NO8
InChi Key: IYGXEHDCSOYNKY-RZHHZEQLSA-N
SMILES: NC(CO)(CO)CO.CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)C[C@H](O)[C@@H]1C\C=C/CCCC(O)=O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results