accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 38454-28-3

IUPAC Name: 4-(hexyloxy)phenyl 4-butylbenzoate
Synonyms: 4-(hexyloxy)phenyl 4-butylbenzoate
Molecular Weight (g/mol): 354.49
Molecular Formula: C23H30O3
InChi Key: QZYDWVAYBOXUCW-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(OC(=O)C2=CC=C(CCCC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results