accessibility menu, dialog, popup

UserName

CAS RN 38291-82-6

IUPAC Name: pentanehydrazide
Synonyms: valeric acid hydrazide
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: PJBQYCIDGYKEMN-UHFFFAOYSA-N
SMILES: CCCCC(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results