accessibility menu, dialog, popup

UserName

H3CNHSNHNNH2NH2HCl

CAS RN 381670-29-7

IUPAC Name: 1-[(diaminomethylidene)amino]-3-ethylthiourea hydrochloride
Synonyms: 1-[(diaminomethylidene)amino]-3-ethylthiourea hydrochloride
Molecular Weight (g/mol): 197.69
Molecular Formula: C4H12ClN5S
InChi Key: LLSXLCPLOKTRCV-UHFFFAOYSA-N
SMILES: Cl.CCNC(=S)NN=C(N)N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results